Anti-HAVCR1 Rabbit Polyclonal Antibody
Catalog # BSBTRP1093
Fournisseur: Boster Bio
Spécifications
- Antibody type:Primary
- Antigen name:hepatitis A virus cellular receptor 1
- Antigen symbol:HAVCR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:26762
- Antigen synonyms:TIM1|TIM-1|KIM-1|CD365|HAVCR|HAVCR-1|TIMD1|TIMD-1|TIM|KIM1
- Size:100 µg
- Pk:100 µG
Spécifications
À propos de cet article
Rabbit IgG polyclonal antibody for Hepatitis A virus cellular receptor 1 homolog(HAVcr-1)(HAVCR1) detection. Tested with WB in Human.
Type: Primary
Antigen: HAVCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human