Glycofural, Sigma-Aldrich®
: Merck
:31692-85-0
:
:C₇H₁₄O₃
:MFCD00801146
:146.186 g/mol
:M-Clarity™ quality level = MQ200
- Solubility:(20 °C) soluble
- Boiling Point:176 °C (40 hPa)
- Refractive Index:1.458 (27 °C, 589 nm)
- Density:1.08 g/cm3 (20 °C)
- Storage temperature:+15...+25 °C
Frequently Bought Together