Precision hydrometers, ASTM E100
Supplier: Ludwig Schneider GmbH & CO. KG
Certificates
About this item
These precision ASTM E100 hydrometers are designed for specific gravity or relative density.
- Reference temperature: 15,5 °C (60/60 °F)
- Without thermometer
- Suitable for calibration
Ordering information: Available with either a works or a DAkkS calibration certificate on request
Packaging: Each packed in a square plastic sleeve.
Specifications
- Catalog No:
- SCNL3222023
- SCNL3222002
- SCNL3222016
- SCNL3222017
- SCNL3222057
- SCNL3222015
- SCNL3222020
- SCNL3222013
- SCNL3222018
- SCNL3222014
- SCNL3222019
- SCNL3222001
- SCNL3222059
- SCNL3222056
- SCNL3222058
- SCNL3222005
- SCNL3222012
- SCNL3222024
- SCNL3222006
- SCNL3222003
- SCNL3222022
- SCNL3222004
- SCNL3222027
- SCNL3222055
- SCNL3222051
- SCNL3222054
- SCNL3222025
- SCNL3222007
- SCNL3222011
- SCNL3222026
- SCNL3222021
- SCNL3222052
- SCNL3222053
- Reference temperature:
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C
- 15,5 °C