Orders LinkContactUsLinkComponent
Separating funnel, pear shaped
Separating funnel, pear shaped
Supplier:   SMITH SCIENTIFIC
Separating funnel, pear shaped
Supplier:  SMITH SCIENTIFIC
Certificates
Separating funnel, pear shaped
2 Options Available Below
Product Details & Documents
Borosilicate glass 3.3, with plastic stopper.
  • Stopcock with glass key
  • Very good temperature resistance
  • Very good chemical resistance
Product Family Options
Glass key
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 250 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,70 €
    95,70 €
    1 * 1 ST
       95,70 €
PTFE key
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 250 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €