Dust test kit
Dust test kit
Catalog # SIMESP3200
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Dust test kit allows assessment of the quantity and size of dust particles on surfaces prepared for painting.
- Report forms for five test results to be filled in pre-defined areas
- Comparator display board to assess dust quantity and particle size
- Sample transfer sheet in resistant and transparent plastic with pre-printed positions
Used either as a pass or fail test or event as a permanent record of the presence of dust.
Certificeringen: ISO 8502-3
Levering: Supplied with two AA batteries, illuminated magnifier, adhesive tape, pair of scissors, pictorial manual, comparator board, transfer sheet and report forms.
Documents
TQC Company Video
Video 's
Specifications
- Description:Dust test kit
Accessories
Product Information
- Catalog #AvailabilityPrice
- Display board (×2)
Product Information
- Catalog #AvailabilityPrice
- Standardized tape
Product Information
- Catalog #AvailabilityPrice
- Report forms
Product Information
- Catalog #AvailabilityPrice
- Sample transfer sheet (×10)
Product Information
- Catalog #AvailabilityPrice
- Spring loaded roller in titanium anodised aluminumSpring loaded roller in titanium anodised aluminumSIMESP3600Log in for availability1 * 1 ST - 522,00 €