Tapes for adhesion test
About this item
This tape consists of 23 microns polyester films backing for improved moisture resistance, weight and space savings. These tapes are ideal to perform a cross cut hatch adhesion test.
- Outstanding dielectric strength
- Resistant to solvents, freon and transformer oils
- Good peel strength to check paints and varnishes
- Temperature resistance up to 130 °C, and classified under thermal class B
Different lengths available.
Colour: Transparent
Certifications: Complies with ISO 2409:2003 or EN ISO 2409:1999.
4 Options Available Below
Product Details & Documents
This tape consists of 23 microns polyester films backing for improved moisture resistance, weight and space savings. These tapes are ideal to perform a cross cut hatch adhesion test.
- Outstanding dielectric strength
- Resistant to solvents, freon and transformer oils
- Good peel strength to check paints and varnishes
- Temperature resistance up to 130 °C, and classified under thermal class B
Different lengths available.
Colour: Transparent
Certifications: Complies with ISO 2409:2003 or EN ISO 2409:1999.
Documents
Product Family Options
Product Information
- WidthAvailabilityPrice
- SIMESP3020Tape for cross cut adhesion test, glass reinforced, 1 roll of 10,0 m, acc. to EN ISO 2409:1999Tape for cross cut adhesion test, glass reinforced, 1 roll of 10,0 m, acc. to EN ISO 2409:199925,0 mmLog in for availability1 * 1 ST42,70 €
- SIMESP3007Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:200325,0 mmLog in for availability1 * 1 ST25,70 €
- SIMESP3010Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:200325,0 mmLog in for availability1 * 3 ROU37,90 €
- SIMESP3025Tape for cross cut adhesion test, glass reinforced, 1 roll of 50,0 m, acc. to EN ISO 2409:1999Tape for cross cut adhesion test, glass reinforced, 1 roll of 50,0 m, acc. to EN ISO 2409:199925,0 mmLog in for availability1 * 1 ST61,00 €