Orders LinkContactUsLinkComponent
Precision thermometers, low temperature
Precision thermometers, low temperature
Supplier:   Ludwig Schneider
Precision thermometers, low temperature
Supplier:  Ludwig Schneider
Certificates
Precision thermometers, low temperature
Precision thermometers, low temperature
11 Options Available Below
Product Details & Documents
These precision, low temperature laboratory thermometers are suitable for official certification.
  • Total immersion
  • Colourless prism or yellow back round capillary
Pentane, alcohol or toluene, enlcosed or stem type.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Ordering information: Available with works certificate or with DAkkS calibration certificate (optional).
Product Family Options
Enclosed type
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1042402
    Enclosed type, with blue reflecting prismatic capillary form
    Enclosed type, with blue reflecting prismatic capillary form
    −150...+30 °C
    1,0 °C
    300 mm
    Pentane
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,60 €
    72,60 €
    1 * 1 ST
       72,60 €
  • SCNL1042403
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −100...+100 °C
    1,0 °C
    300 mm
    ALCH
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,00 €
    72,00 €
    1 * 1 ST
       72,00 €
  • SCNL1042405
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −80...+30 °C
    1,0 °C
    300 mm
    Toluol
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,30 €
    47,30 €
    1 * 1 ST
       47,30 €
  • SCNL1042504
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −50...+50 °C
    0,5 °C
    300 mm
    Toluol
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,30 €
    47,30 €
    1 * 1 ST
       47,30 €
  • SCNL1042602
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −50...+50 °C
    0,2 °C
    420 mm
    Toluo
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,60 €
    77,60 €
    1 * 1 ST
       77,60 €
Stem type
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1052006
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −50...+50 °C
    1,0 °C
    280 mm
    Toluol
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,30 €
    37,30 €
    1 * 1 ST
       37,30 €
  • SCNL1052009
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −50...+30 °C
    1,0 °C
    280 mm
    Toluol
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,20 €
    36,20 €
    1 * 1 ST
       36,20 €
  • SCNL1052003
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −100...+100 °C
    1,0 °C
    300 mm
    ALCH
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,70 €
    64,70 €
    1 * 1 ST
       64,70 €
  • SCNL1052004
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −100...+30 °C
    1,0 °C
    300 mm
    ALCH
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,70 €
    46,70 €
    1 * 1 ST
       46,70 €
  • SCNL1052005
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −80...+30 °C
    1,0 °C
    300 mm
    Toluol
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €
  • SCNL1052104
    Stem type, with yellow back round capillary form
    Stem type, with yellow back round capillary form
    −50...+50 °C
    0,5 °C
    300 mm
    Toluol
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,10 €
    42,10 €
    1 * 1 ST
       42,10 €