ContactUsLinkComponent
Surface profile and coating thickness gauge
Surface profile and coating thickness gauge
Catalog # SIMESP1560
Supplier:  Industrial Physics
undefined
Surface profile and coating thickness gauge
Catalog # SIMESP1560
Supplier:  Industrial Physics
Specifications
  • Range:
    0 - 3,4 mm
  • Description:
    Combined roughness and thickness gauge
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Combination gauge equipped with two different tips: Sharp needle for surface roughness and round tip for coating thickness.
  • Sharp tungsten carbide replaceable tip for surface profile measurement
  • Hardened round tip for coating thickness measurement
  • Glass plate for zero calibration and a leather carry pouch
Certifications: ISO 2808, ASTM D4417-B, JIS K 5600-1-7, BS EN ISO 2808, BS 3900-C5, IMO MSC 215(82).
Delivery: Supplied with two tips and a glass calibration plate in a leather pouch.