ContactUsLinkComponent
Ultra-Plex™ 1-Step ToughMix® (4X)
Ultra-Plex™ 1-Step ToughMix® (4X)
Supplier:  Quantabio
Certificates
undefined
Ultra-Plex™ 1-Step ToughMix® (4X)
Supplier:  Quantabio
Certificates
Specifications
  • Application:
    RT-qPCR probe
  • Inhibitor-rich sample:
    Yes
  • Multiplexing:
    Yes
  • One step:
    Yes