ContactUsLinkComponent
Wireless temperature and humidity data loggers, AiroSensor
Wireless temperature and humidity data loggers, AiroSensor
Supplier:  SenseAnywhere
Certificates
undefined
Wireless temperature and humidity data loggers, AiroSensor
Supplier:  SenseAnywhere
Specifications
  • Catalog No:
  • SENA0630125977842
  • SENA0630125977859
  • 620-2402
  • 620-2404
  • Model:
  • AiroSensor
  • AiroSensor ER
  • Interface:
  • WiFi
  • WiFi
  • Protection class:
  • IP 62
  • IP 62
  • Battery life:
  • Up to 10 years with 5 min interval = 1 million events
  • Up to 10 years with 5 min interval = 1 million events
  • W×D×H:
  • 41×24×33 mm
  • 131×24×33 mm
  • Data memory - No. of readings:
  • 15000 events internally (52 days @ 5 min)
  • 15000 events internally (52 days @ 5 min)
  • Batteries:
  • Built-in
  • Built-in
Specifications
Related Information
AccessPoint
Product Information
Frequently Bought Together