Order Entry
Belgium
ContactUsLinkComponent
Adapter, BNC connector to 4 mm jack plugs
Adapter, BNC connector to 4 mm jack plugs
  BINH1002750
undefined
Adapter, BNC connector to 4 mm jack plugs
  BINH1002750

 

  • Description:
    Adapter, BNC connector to 4 mm Jack plugs with 19 mm spacing.
  • Pk:
    1