Solid phase extraction cartridges, Resprep®, method specific
Supplier: Restek
Jump to:
About this item
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
5 Options Available Below
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Product Family Options
Product Information
- Column hardwareCapacityBed weightAvailabilityPrice
- REST25499Ultra Quat SPE method, for use in HPLC analysis of paraquat/diquatUltra Quat SPE method, for use in HPLC analysis of paraquat/diquatPP6 ml500 mgLog in for availability1 * 30 ST317,00 €
- REST26032EPA methods 521 and 522, for use in EPA method 521 for nitrosamines in drinking water and EPA method 522 for 1,4-dioxane in drinking water, with activated charcoal packingEPA methods 521 and 522, for use in EPA method 521 for nitrosamines in drinking water and EPA method 522 for 1,4-dioxane in drinking water, with activated charcoal packingPP6 ml2 gLog in for availability1 * 30 ST360,00 €
- REST26063EPA method 548.1, for extraction of endothall from aqueous samplesEPA method 548.1, for extraction of endothall from aqueous samplesPP6 ml-Log in for availability1 * 30 ST1 280,00 €
- REST26093RDX method, for extraction of explosive compounds from water samplesRDX method, for extraction of explosive compounds from water samplesPP6 ml500 mgLog in for availability1 * 30 ST875,00 €
- REST24049Organo tin method, for high capacity clean-up of butyl and phenyl tin compounds from soil, water and biota. Mixed bedOrgano tin method, for high capacity clean-up of butyl and phenyl tin compounds from soil, water and biota. Mixed bedPP60 ml-Log in for availability1 * 16 ST463,00 €
This product has been blocked by your organization. Please contact your purchasing department for more information
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.