Orders LinkContactUsLinkComponent
Forceps set
Forceps set
Set of four forceps
Catalog # 232-0187
Supplier:   RSG
Forceps set
Catalog # 232-0187
Supplier:  RSG
Forceps set
Forceps set
Set of four forceps
Price
66,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Set contains four pairs of forceps made from stainless steel:
1× fine pointed
1× blunt ended
1× fine with guide pin
1× cover slip
Guidebook and case
Specifications
  • Description:
    Set of four forceps