Avantor® ACE® C8-300, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® C8-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
- Designed for peptide and protein separations
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Specifications
- Catalog No:
- ACE-222-1510
- ACE-222-1520
- ACE-222-0510
- ACE-222-0520
- ACE-222-1502
- ACE-222-1501
- ACE-222-2510
- ACE-222-2520
- ACE-222-2504
- ACE-222-1220
- ACE-222-0103GD
- ACE-222-2503
- ACE-222-0346
- ACE-222-0110GD
- ACE-222-2546
- ACE-222-1204
- ACE-222-0302
- ACE-222-1020
- ACE-222-1046
- ACE-222-1010
- ACE-222-1002
- ACE-222-1546
- ACE-222-7530
- ACE-222-7520
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- 5,0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar
- 400 bar
- 400 bar
- 400 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 400 bar
- 400 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 400 bar
- 275 bar
- 400 bar
- 275 bar
- 275 bar
- 275 bar
- 400 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- USP code:
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Guard holders and accessories
Product Information
Frequently Bought Together