Capillary columns, Rxi®-5ms (fused silica)
: Restek
New Product
General purpose columns for semivolatiles, phenols, amines, residual solvents, drugs of abuse, pesticides, PCB congeners (e.g. Aroclor mixes), solvent impurities.
Low polarity phase; Crossbond® 100% diphenyl dimethyl polysiloxane
Inert column, tested and guaranteed for ultra-low bleed, improved signal-to-noise ratio for better sensitivity and mass spectral integrity.
Temperature range: −60 to 330/350 °C
Similar phases: HP-1 ms, HP-1 msUI, DB-1 ms, DB-1 msUI, Ultra-1, VF-1 ms, ZB-1, ZB-1 ms.
Equivalent to USP G1, G2 and G38 phases.
- Catalog No:
- REST13402
- REST13409
- REST13439
- REST13421
- REST13423
- JJBEI13438
- REST13424
- REST13438
- REST13481
- REST13420
- REST13455
- REST13453
- REST13426
- REST13442
- Temperature limit:
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
- −60...330/350 °C
Frequently Bought Together