Inserts
: MIELE
- Catalog No:
- 141-0492
- 141-0335
- MIEL2648950
- MIEL3830310
- 141-0518
- MIEL3203530
- MIEL3203500
- 141-0525
- MIEL5043640
- MIEL5109190
- 141-0412
- 141-0471
- 141-0470
- MIEL5649700
- 141-0439
- 141-0423
- 141-0330
- 141-0296
- 141-0352
- 141-0353
- 141-0297
- 141-0354
- 141-0298
- MIEL3808500
- MIEL3808510
- MIEL3808520
- MIEL3808550
- 141-0299
- 141-0356
- MIEL3808530
- 141-0479
- 141-0379
- 141-0355
- 141-0380
- 141-0381
- 141-0387
- 141-0388
- 141-1291
- MIEL12897190
- MIEL12669750
- MIEL12531630
- MIEL11017170
- W×D×H:
- 543×473×1 mm
- 234×459×221 mm
- 180x445x180 mm
- 255×230×55 mm
- 418×471×235 mm
- 460×460×110 mm
- 180×445×110 mm
- 150×445×40 mm
- 250×170×76 mm
- 150×470×480 mm
- 530×53×300 mm
- 357×522×155 mm
- 357×522×173 mm
- 220×450×50 mm
- 363×533×122 mm
- 460×460×148 mm
- 460×460×224 mm
- 445×445×83 mm
- 445×445×113 mm
- 445×445×102 mm
- 357×522×185 mm
- 150×225×55 mm
- 558×352×55 mm
- 178×279×157 mm
- 492×503×118 mm
Product Family Options
Inserts
- DescriptionForW×D×HW×D×H ext.
- A 7/1 insert, perforated plate, for E 775/1, E 735/1 and E 701/1
7×7 mm perforations, 3 mm ridgeG 7825, G 7826543×473×1 mm-Specifications
Cat. No.141-0492W×D×H543×473×1 mm - A 12/1 Insert for the gentle cleaning of delicate items by interrupting the spray jet, with perforations 7×7 mm (224×429×60 mm)G 7804, G 7883, G 7893, G 7883 CD, G 7835 CD, G 7836 CD234×459×221 mm1 * 1 ST77,90 €
Specifications
Cat. No.141-0335W×D×H234×459×221 mm - E 130 insert with 11 brackets, for 10 tabletsPG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592180x445x180 mm1 * 1 ST
Specifications
Cat. No.MIEL2648950W×D×H180x445x180 mm - E 143, insert 1/4, mesh tray, for various items, with 2 hinged handles, 5 mm mesh spacing, 5 mm frame all roundPG 8535/36, PG 8582, PG 8582 CD, PG 8592255×230×55 mm1 * 1 ST
Specifications
Cat. No.MIEL3830310W×D×H255×230×55 mm - E 319/3 Surface filter for coarse wasteG 7804, G 7883/93, G 7883 CD, G 7835/36 CD418×471×235 mm1 * 1 ST300,20 €
Specifications
Cat. No.141-0518W×D×H418×471×235 mm - E 378, insert 1/1 for a range of different utensils, with 2 carrying handles, for use in lower basket
1,7 mm mesh spacing, 5 mm frame all roundG 7882, G 7892, G 7882 CD, G 7835/36 CD460×460×110 mm1 * 1 ST427,27 €Specifications
Cat. No.MIEL3203530W×D×H460×460×110 mm - E 379, insert 1/2 for various utensils, upper and lower baskets, with 2 handles, 0,8 mm gauge wire, 5 mm frame all roundG 7826, G 7831, PG 8528, PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592180×445×110 mm1 * 1 ST308,20 €
Specifications
Cat. No.MIEL3203500W×D×H180×445×110 mm - E 430/1 insert, 1/3 mesh tray, for various instruments, wire gauge with mesh size 5 mm, for upper and lower basketG 7881, G 7891150×445×40 mm1 * 1 ST
Specifications
Cat. No.141-0525W×D×H150×445×40 mm - E 468, insert 1/4 mesh basket with lid and divisions, for various utensils, stainless steel welded meshG 7882, G 7892, G 7882 CD, G 7835/36 CD250×170×76 mm1 * 1 ST267,90 €
Specifications
Cat. No.MIEL5043640W×D×H250×170×76 mm - E 484 1/1 insert, wire mesh for various utensils, 8 mm mesh spacingG 7823, G 7824, G 7825, G 7826, PG 8528, PG 8535/36, PG 8582, PG 8582 CD, PG 8592150×470×480 mm-
Specifications
Cat. No.MIEL5109190W×D×H150×470×480 mm - E 500 insert for soldering frames, for lower basket U 874/1, for holding 8 solder frames, 8 compartments, distance 53 mm (530×545×400 mm)Glassware washing machines530×53×300 mm-
Specifications
Cat. No.141-0412W×D×H530×53×300 mm - E 963 insert, with 33×3 holders for beakers up to 250 mlG 7827357×522×155 mm1 * 1 ST1 213,79 €
Specifications
Cat. No.141-0471W×D×H357×522×155 mm - E 965 insert, with 15×3 holders for beakers 250–600 mlG 7827357×522×173 mm-
Specifications
Cat. No.141-0470W×D×H357×522×173 mm - Insert 1/2, E523 for sieve trays, 7 bracketsPG 8581, PG 8591220×450×50 mm1 * 1 ST266,95 €
Specifications
Cat. No.MIEL5649700W×D×H220×450×50 mm - Mesh tray E 969, for various items, for E 900-4, E 935-3, E 975-2 and E 941 with 945
Perforated metal plate: 7×7×3 mmG 85727/28363×533×122 mm1 * 1 ST205,20 €Specifications
Cat. No.141-0439W×D×H363×533×122 mm - Retaining frame E 750 for insert E 106-111Washer-disinfectors-
Specifications
Cat. No.141-0423W×D×H
Inserts for test tubes, flasks and wide-mouthed glassware
- DescriptionForW×D×HW×D×H ext.
- AK 12 Insert suitable for breast milk pumps, beakers and funnels1 * 1 ST114,00 €
Specifications
Cat. No.141-0330W×D×H - E 106 Insert for wide neck flasks and measuring cylinders, equipped with 10 spring hooks ×175 mm height and 16×105 mm height1 * 1 ST181,45 €
Specifications
Cat. No.141-0296W×D×H - E 106/1 Insert for wide neck flasks and measuring cylinders, equipped with 26 spring hooks ×105 mm height1 * 1 ST181,45 €
Specifications
Cat. No.141-0352W×D×H - E 106/2 Insert for wide neck flasks and measuring cylinders, equipped with 13 spring hooks ×175 mm height1 * 1 ST145,35 €
Specifications
Cat. No.141-0353W×D×H - E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars1 * 1 ST267,90 €
Specifications
Cat. No.141-0297W×D×H - E 110 Insert suitable for 10 glass beakers between 250 - 600 ml, equipped with 10×3 stay bars1 * 1 ST196,65 €
Specifications
Cat. No.141-0354W×D×H - E 111 Insert suitable for 8 glass beakers between 600 - 1000 ml, equipped with 8×3 stay bars1 * 1 ST196,65 €
Specifications
Cat. No.141-0298W×D×H - E 124-06 insert for infusion bottles, 16 fields, square measure 100x100 mmPG 8535/36, PG 8582, PG 8582 CD, PG 8592460×460×148 mm1 * 1 ST
Specifications
Cat. No.MIEL3808500W×D×H460×460×148 mm - E 125-06 insert for infusion bottles, 9 fields, square measure 125×125 mmPG 8535/36, PG 8582, PG 8582 CD, PG 8592460×460×224 mm-
Specifications
Cat. No.MIEL3808510W×D×H460×460×224 mm - E 126-06 insert for infusion bottles, 48 fields, square measure 45×45 mmPG 8535/36, PG 8582, PG 8582 CD, PG 8592445×445×83 mm-
Specifications
Cat. No.MIEL3808520W×D×H445×445×83 mm - E 129-06 insert for infusion bottles, 20 fields, square measure 84×84 mmPG 8535/36445×445×113 mm-
Specifications
Cat. No.MIEL3808550W×D×H445×445×113 mm - E 144 Insert suitable for 18 glass beakers, equipped with 18×3 stay bars, W×D×H: 200x445x131 mm1 * 1 ST295,45 €
Specifications
Cat. No.141-0299W×D×H - E 149 Insert suitable for 80 test tubes up to 16×105 mm, 80 Compartments at 18×18 mm and additionally equipped with a lid, W×D×H: 528×527×206 mm1 * 1 ST484,50 €
Specifications
Cat. No.141-0356W×D×H - Insert 1/1 E 127 for 44 flasks 100 mlPG 8535/36, PG 8582, PG 8582 CD, PG 8592445×445×102 mm-
Specifications
Cat. No.MIEL3808530W×D×H445×445×102 mm - Insert E 960/1, with 20 large and 26 small spring hook for wide-necked Erlenmeyer flask and measuring cylinderG 7827357×522×185 mm-
Specifications
Cat. No.141-0479W×D×H357×522×185 mm
Inserts for Petri dishes, slides and watch glasses
- DescriptionForW×D×HW×D×H ext.
- E 118 suitable for 38 half petri dishes with Ø 100 mm, equipped with 38 holders, height 70 mm, distance approx. 26 mm1 * 1 ST377,15 €
Specifications
Cat. No.141-0379W×D×H - E 134 Insert for 210 slides, equipped with 26 compartments at 26×11 mm, wire thickness 3 mm, W×D×H: 525×528×141 mm1 * 1 ST315,40 €
Specifications
Cat. No.141-0355W×D×H - E 136 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm1 * 1 ST436,05 €
Specifications
Cat. No.141-0380W×D×H - E 137 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm1 * 1 ST436,05 €
Specifications
Cat. No.141-0381W×D×H - E 402 Insert suitable for 44 watch glasses Ø 80 - 125 mm, equipped with 23 bars spaced 15 mm apart, W×D×H: 600×805×1003 mm1 * 1 ST329,65 €
Specifications
Cat. No.141-0387W×D×H - E 403 Insert suitable for 105 watch glasses Ø 50 - 60 mm, equipped with 36 bars spaced 9 mm apart1 * 1 ST404,70 €
Specifications
Cat. No.141-0388W×D×H
Inserts for GYN and ENT instruments
- DescriptionForW×D×HW×D×H ext.
- E 363 insert with lid, for upper and lower baskets, for GYN and ENT instrumentsG 7831, PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592150×225×55 mm1 * 1 ST369,55 €
Specifications
Cat. No.141-1291W×D×H150×225×55 mm - E945/2 module for inserts - holding frame for inserts 558×352×55 mm (W×D×H); Compatible with PG 8527/28558×352×55 mm1 * 1 STNew Product
Specifications
Cat. No.MIEL12897190W×D×H558×352×55 mmNew Product - E 416 insert 178×279×157 mm W×D×H for optimum loading of 6 single and two-piece specula, 7 holders 40 mm apart, for use in upper (only with singlepart specula) and lower basket; For use with G 7831, PG 8535/36/62/82/92, PWD 8534, PWD 8682/92178×279×157 mm1 * 1 STNew Product
Specifications
Cat. No.MIEL12669750W×D×H178×279×157 mmNew Product - A 315/1 module 492×503×118 mm (W×D×H) for increased capacity and reprocessing of lumened instruments, increases capacity from 22 to 44, can be used in all A 151 lower baskets available on the market, compatible with all standard dental and ENT accessories; Injector nozzles not included, A 800 tubular filter for use in either; Clearances in lower basket A 151: H 210 ±30, W 383, D 479 mm injector manifold (not included); For use with: PG 8581/62/82/91/92, PWA 8672, PWD 8682/92492×503×118 mm1 * 1 STNew Product
Specifications
Cat. No.MIEL12531630W×D×H492×503×118 mmNew Product
Inserts for APWD 063 Trays or Cassettes
- DescriptionForW×D×HW×D×H ext.
- Insert for the optimum loading of up to 8 trays or cassettes, equipped with 9 holders, spacing: 40 mmPWD 8531, PWD 8532103×389×110 mm1 * 1 ST
Specifications
Cat. No.MIEL11017170W×D×H