Order Entry
Belgium
ContactUsLinkComponent
Ion exchange chromatography media, Ion Exchanger I
Ion exchange chromatography media, Ion Exchanger I
Catalog # 1.04765.0500
Supplier:  Merck
CAS Number:  
undefined
Ion exchange chromatography media, Ion Exchanger I
Catalog # 1.04765.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic Bottle
  • Particle Size:
    0.3-1.2 mm
  • Pk:
    500 g
  • Description:
    Ion exchanger I
  • Seq:
    01
  • Type:
    Solid

Specifications

About this item

Ion Exchanger I is a strong acid cation exchangers widely used in separation of noble earths and amino acids, water treatment, analysis and food industry. The bond strength of equivalent ions increases with decreasing diameter of the hydrated ion.

Maximum working temperature: 120 °C
Exchange capacity: >1,7 mval/ml