Orders LinkContactUsLinkComponent
Pipette tips, FlexiBulk, Optifit
Pipette tips, FlexiBulk, Optifit
   Sustainable Options Available
Supplier:   Sartorius
Pipette tips, FlexiBulk, Optifit
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Pipette tips, FlexiBulk, Optifit
5 Options Available Below
Product Details & Documents
The FlexiBulk concept combines the low cost of bulk tips with saving space, improving usability and securing tip purity.
  • Reduces up to 40% on pack volume; cuts packaging waste by nearly 50%
  • Saves time and effort: Well-organized FlexiBulk pack makes tip picking quicker and more convenient
  • Tips are certified as free of RNase, DNase and endotoxins
  • Tips are manufactured under ISO Class 8 cleanroom conditions
  • Lid provides an airtight seal during transport and storage for secured purity
  • Lid is easy to close between uses to protect the tips from dust
FlexiBulk is small-sized and made of durable PET (polyethylene terephthalate) material that can be used and stacked as is – without any outer cardboard box needed. The orderly arrangement of tips allows the FlexiBulk tip packs to be approximately 40% smaller than conventional bulk tip cartons, where tips are packed in random order.
Certifications: Compliance with ISO 9001, ISO 14001 and ISO 13485 quality standards.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-3887
  • 613-3888
  • 613-3890
  • 613-3889
  • 613-3891
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
Specifications
Product Family Options
Product Information
  • Tip style
    Volume
    Length
    Packed
    Availability
    Price
  • FlexiBulk
    0,5 - 200 µl
    51,0 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 38,50 €
    38,50 €
    1 * 960 ST
       38,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-3887
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • FlexiBulk
    5 - 350 µl
    54,0 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 43,60 €
    43,60 €
    1 * 960 ST
       43,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-3888
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • FlexiBulk, wide bore
    10 - 1000 µl
    68,5 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 25,70 €
    25,70 €
    1 * 480 ST
       25,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-3890
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • FlexiBulk
    10 - 1000 µl
    71,5 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 20,50 €
    20,50 €
    1 * 480 ST
       20,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-3889
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • FlexiBulk
    50 - 1200 µl
    71,5 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 26,90 €
    26,90 €
    1 * 480 ST
       26,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-3891
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact