Order Entry
Belgium
ContactUsLinkComponent
Ductless filtering fume hoods, Captair® Smart and Midcap 321
Ductless filtering fume hoods, Captair® Smart and Midcap 321
Supplier:  ERLAB CAPTAIRE
New Product
Certificates
undefined
Ductless filtering fume hoods, Captair® Smart and Midcap 321
Supplier:  ERLAB CAPTAIRE
New Product

Specifications

  • Catalog No:
  • ERLAAA602A00011A41
  • ERLAAA602A00011A21
  • ERLAAA606A00011A21
  • ERLAAA602A00011A31
  • Model:
  • Captair® 321 Smart & Midcap
  • Captair® 321 Smart & Midcap
  • Captair® 321 Smart
  • Captair® 321 Smart & Midcap
  • Treated air volume:
  • 200−230
  • 200−230
  • 200−230
  • 200−230
  • Air velocity at the front panel:
  • 0,4 - 0,6 m/s
  • 0,4 - 0,6 m/s
  • 0,4 - 0,6 m/s
  • 0,4 - 0,6 m/s
  • Lighting:
  • LED > 650 lux
  • LED > 650 lux
  • LED > 650 lux
  • LED > 650 lux
  • Power:
  • 60 W
  • 60 W
  • 60 W
  • 60 W
  • Filters:
  • HEPA H14: filtration efficiency: 99,995 % according to MPPS methods, EN1822 standard
    ULPA U17: filtration efficiency 99,999995 % according to MPPS method, EN 1822 standard
  • HEPA H14: filtration efficiency: 99,995 % according to MPPS methods, EN1822 standard
    ULPA U17: filtration efficiency 99,999995 % according to MPPS method, EN 1822 standard
  • HEPA H14: filtration efficiency: 99,995 % according to MPPS methods, EN1822 standard
    ULPA U17: filtration efficiency 99,999995 % according to MPPS method, EN 1822 standard
  • HEPA H14: filtration efficiency: 99,995 % according to MPPS methods, EN1822 standard
    ULPA U17: filtration efficiency 99,999995 % according to MPPS method, EN 1822 standard

Specifications

Related Information

Frequently Bought Together