ContactUsLinkComponent
Shakers, purging method
Shakers, purging method
Catalog # HACHLQV1489900001
Supplier:  Hach
undefined
Shakers, purging method
Catalog # HACHLQV1489900001
Supplier:  Hach
Specifications
  • Shaking motion:
    Rocking/Rotating
  • Description:
    TOC shaker, TOC-X5, for purging method
  • Model:
    TOC-X5
  • Weight:
    2,9 kg
  • W×D×H:
    190×240×135 mm
  • Pk:
    1
Specifications
About this item
Shaker which is used for purging of inorganic carbon (TIC). It has an automatic shutoff system. Accommodates eight cuvettes simultaneously.