Mouse affinity purification resin
Product Details & Documents
ImmunoReagents Mouse IgG Affinity Prep resins are prepared using highly purified species specific immunoglobulins. This resin is suitable for removal of proteins recognising Mouse IgG from liquid solutions, including traditional Immunoabsorptions and Immunoprecipitation protocols.
Immunoglobulins demonstrate a single arc when analysed against normal serum proteins by IEP. The resins provided comprise of immunoglobulins covalently bound to a 4% agarose bead utilising strong and stable hydrazide/aldehyde group chemistry. The Affinity Prep resins have 8 to 10 mg of antibodies per/ml of resin, with a 24 month shelf life reagents.
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.
: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
: ImmunoReagents products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles. Additionally, products can by customized based on specific assay requirements. Contact your Avantor representative for more information.
Specifications
Specifications
- Pk:2 ml
- Description:Purified mouse IgG coupled to 4% agarose beads
- Type:Liquid
- Cat. No.:IMMRIR1013
- Storage temperature:2...8 °C



