Order Entry
Belgium
Orders LinkContactUsLinkComponent
Label printer, BMP™41

Label printer, BMP™41

Supplier:   Brady
Label printer, BMP™41
Supplier:  Brady
Certificates
Label printer, BMP™41

1 Options Available Below

Product Details & Documents

The BMP™41 portable label printer creates die-cut or continuous labels anywhere and at anytime. It can produce up to 25 mm wide labels which can be used as multi area labels mostly on labelling datacom, wires, cables and audio/visual equipments.

Delivery information: Supplied with nickel battery, MC1-1000-595-WT-BK cartridge, USB cable, AC adapter, power cord, product CD, brady printer drivers CD and quick start guide.

Product Family Options

Product Information

  • Print width
    W×D×H
    Weight
    Availability
    Price
  • BRDY133254
    Label printer BMP™41, EU-plug
    Label printer BMP™41, EU-plug
    24,40 mm
    150×90×290 mm
    1,3 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • BRDY711213
    Label printer BMP™41, UK-plug
    Label printer BMP™41, UK-plug
    24,40 mm
    150×90×290 mm
    1,3 kg
    Discontinued
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Quick charger US for M410 label printer, black, 114,30W×69,85H×101,60D mm
    Quick charger US for M410 label printer, black, 114,30W×69,85H×101,60D mm
    BRDY176994
    Log in for availability
    1 * 1 ST - 205,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Rechargeable NiMH battery for M410, BMP41 and BMP61 label printers, grey, 56,90W×109,98×25,00D mm
    Rechargeable NiMH battery for M410, BMP41 and BMP61 label printers, grey, 56,90W×109,98×25,00D mm
    BRDY176993
    Log in for availability
    1 * 1 ST - 83,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • AC adapter for M410, M510, M511, M610 and M611 portable printers, black, 114,30W×31,75H×50,80D mm
    AC adapter for M410, M510, M511, M610 and M611 portable printers, black, 114,30W×31,75H×50,80D mm
    BRDY176990
    BMP41; M410; BMP51; BMP53; M510; BMP61; M610; M611
    Log in for availability
    1 * 1 ST - 124,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.