Temperature control jackets for viscosity cups
Supplier: Industrial Physics
Jump to:
About this item
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
- Adjustable feet for easy leveling
- Built-in spirit level to set jacket exactly horizontal
- Glass plate to prevent dripping before measurement
- Elbow-type quick release couplings
- Titanium anodised aluminium
Delivery information: Supplied with jacket with built-in spirit level, glass plate and tube connection.
2 Options Available Below
Product Details & Documents
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
- Adjustable feet for easy leveling
- Built-in spirit level to set jacket exactly horizontal
- Glass plate to prevent dripping before measurement
- Elbow-type quick release couplings
- Titanium anodised aluminium
Delivery information: Supplied with jacket with built-in spirit level, glass plate and tube connection.
Documents
TQC Company Video
Videos
Product Family Options
Product Information
- AvailabilityPrice
- 769-1131Temperature control jacket with tripod for DIN and ASTM cupsTemperature control jacket with tripod for DIN and ASTM cupsLog in for availability1 * 1 ST569,00 €
- SIMEVF2068Temperature control jacket with tripod for ISO and AFNOR cupsTemperature control jacket with tripod for ISO and AFNOR cupsLog in for availability1 * 1 ST621,00 €