VWR®, Digital Stopwatch
Digital stopwatch, Black/blue, ±0,0005 %, 63,5×19×76,2 mm
Catalog # 609-0212
Supplier: VWR Collection
Jump to:
Product Details & Documents
Quartz-crystal stopwatch with large LCD digits.
- Timing capacity: 9 hours, 59 minutes, 59 seconds and 99 hundredths
- Functions: Single action, time out, cumulative split, interval split, continuous roll-over timing and countdown alarm timing
- Tough, water resistant ABS plastic casing seals out moisture, dust and fumes
- Ideal for enzyme tests, viscosity measurements, animal studies, and all timed lab tests
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with lanyard, battery and certificate.
Specifications
- Colour:Black/blue
- Description:Digital stopwatch
- W×D×H:63,5×19×76,2 mm
- Accuracy:±0,0005 %
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



