Order Entry
Belgium
ContactUsLinkComponent
Qualitative filter papers, grades MN 617 and 617 1/4, circles and folded filters
Qualitative filter papers, grades MN 617 and 617 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Qualitative filter papers, grades MN 617 and 617 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL

Specifications

  • Catalog No:
  • 515-9781
  • MANA534005
  • 512-0508
  • MANA434032
  • MANA534018
  • 512-0506
  • 512-0509
  • 512-0511
  • MANA534011
  • MANA534012
  • 515-9780
  • MANA434024
  • MANA534007
  • 512-0507
  • MANA534009
  • MANA534032
  • 512-0510
  • MANA434015
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • Particle retention:
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • Filtration speed:
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • Ash content:
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%

Specifications

Related Information

Frequently Bought Together