ContactUsLinkComponent
Paint inspection gauge, Super Pig III
Paint inspection gauge, Super Pig III
Catalog # SIMESP1100
Supplier:  Industrial Physics
undefined
Paint inspection gauge, Super Pig III
Catalog # SIMESP1100
Supplier:  Industrial Physics
Specifications
  • Model:
    SuperPig III
  • Description:
    Paint inspection gauge
  • Pk:
    1
Specifications
About this item
This inspection gauge is a destructive precision tool for inspection and thickness measurement, on single or multiple coats, on virtually all substrates, including wood, plastics, metals.
  • Titanium anodised aluminum
  • Three knife sizes and one crosscut knife in one holder
  • Ergonomic design with revolving system
  • Bright white LED ensure clear vision through the microscope
  • Small size eases use in corners
  • Engraved scaling (D-factor) for easy calculations
Observes and measures substrate and film defects. It is a very stable instrument, also the integrated microscope has an excellent focus.
The microscope is provided with a double scale (mm and inch) which allows you to calculate to micrometers and mils.
Delivery: Supplied with black marker, cutters (1, 2, 4 ), hexadiagonal wrench and black leather case with belt clip.