ContactUsLinkComponent
Gas detection tubes for mercaptans
Gas detection tubes for mercaptans
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for mercaptans
Supplier:  Dräger
Specifications
  • Catalog No:
  • 151-0160
  • 118-0322
  • 151-0182
  • Model:
  • Mercaptan 0.5/a
  • Mercaptan 0.1/a
  • Mercaptan 20/a
  • Colour change:
  • White to yellow
  • Yellow to red
  • White to yellow brown
  • Humidity:
  • 3 - 15 mg/l H₂O
  • 2 - 40 mg/l H₂O
  • 3 - 30 mg/l H₂O
  • Measurement time:
  • 8 minutes
  • 3 minutes/40 seconds
  • 2,5 minutes
  • No. of strokes:
  • 20
  • 10 / 2
  • 10
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 0,5 - 5 ppm
  • 0,1 - 2,5 / 3 - 15 ppm
  • 2 - 100 ppm
  • Temperature range:
  • 10…40 °C
  • 5…40 °C
  • 0…50 °C