ContactUsLinkComponent
Gas detection tubes for ethylene oxide
Gas detection tubes for ethylene oxide
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for ethylene oxide
Supplier:  Dräger
Specifications
  • Catalog No:
  • 151-0108
  • 118-0292
  • Model:
  • Ethylene oxide 25/a
  • Ethylene oxide 1/a
  • Colour change:
  • Pale yellow to turquoise green
  • White to pink
  • Humidity:
  • 3 - 15 mg/l H₂O
  • 3 - 15 mg/l H₂O
  • Measurement time:
  • 6 minutes
  • 8 minutes
  • No. of strokes:
  • 30
  • 20
  • Standard deviation:
  • ±20 - 30 %
  • ±20 - 30 %
  • Standard range:
  • 25 - 500 ppm
  • 1 - 15 ppm
  • Temperature range:
  • 15…30 °C
  • 10…30 °C