ContactUsLinkComponent
Gas detection tubes for toluene
Gas detection tubes for toluene
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for toluene
Supplier:  Dräger
Specifications
  • Catalog No:
  • 118-0315
  • 118-0316
  • 118-0313
  • Model:
  • Toluene 50/a
  • Toluene 100/a
  • Toluene 5/b
  • Colour change:
  • White to brown
  • White to brown violet
  • White to pale brown
  • Humidity:
  • 5 - 12 mg/l H₂O
  • ≤ 30 mg/l H₂O
  • ≤ 20 mg/l H₂O
  • Measurement time:
  • 1,5 minutes
  • 1,5 minutes
  • 2/10 minutes
  • No. of strokes:
  • 5
  • 10
  • 2/10
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 50 - 400 ppm
  • 100 - 1800 ppm
  • 50 - 300 / 5 - 80 ppm
  • Temperature range:
  • 0…30 °C
  • 0…40 °C
  • 2…40 °C