Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microplates
PCR Plates
PCR plates, 384-well, with full skirt, Multiply®
PCR plates, 384-well, with full skirt, Multiply®
Catalog # SARS72.1984.202
Supplier:
SARSTEDT
Expand image
PCR plates, 384-well, with full skirt, Multiply®
Catalog # SARS72.1984.202
Supplier:
SARSTEDT
Specifications
No. of wells:
384
Profile:
Standard
Skirt:
Fully skirted
Well colour:
Clear
Environmentally Preferable:
Pk:
50
Well volume:
40 µl
Specifications
About this item
Product Details
Product Details