ContactUsLinkComponent
PCR plates, 96-well, without skirt, Multiply®
PCR plates, 96-well, without skirt, Multiply®
Catalog # SARS72.1978.202
Supplier:  SARSTEDT
undefined
PCR plates, 96-well, without skirt, Multiply®
Catalog # SARS72.1978.202
Supplier:  SARSTEDT
Specifications
  • No. of wells:
    96
  • Profile:
    Standard
  • Skirt:
    Non-skirted
  • Well colour:
    Clear
  • Description:
    PCR plate, with raised well rims
  • Environmentally Preferable:
  • Pk:
    100