Orders LinkContactUsLinkComponent
Label printer BBP™12
 :   Brady
Label printer BBP™12
 :  Brady
Label printer BBP™12
Label printer BBP™12
The BBP™12 bench top label printer is a compact, reliable and affordable entry level printer to identify cables, components, products and laboratory samples for customers who require low volumes of various, quality identification labels. The printer can print on a multitude of materials in various sizes including nylon cloth, paper, polyester, polyimides, polyolefin, polypropylene, tamper evident materials, vinyl (also self-laminating), vinyl cloth, to produce labels, tags, cable markers, sleeves (both ladder and continuous).
  • Accurate image printing, 300 dpi gives good print resolution
  • Supports a multitude of materials from 10 to 112 mm wide
  • Easy to use with intuitive software interface and full colour LCD display
  • Can work in a network or stand-alone
  • Print speed = 100 mm/sec
  • Real time clock feature
 : The LAB-KIT comprises the BBP12 label printer with an unwinder (media holder), USB cable, power cord, product CD, Quick Start Guide, a LabelMark Pro CD.
Product Family Options
  • Description
  • BBP™12 Desktop label printer, 300 dpi, English version with UK-plug
  • BBP™12 Desktop label printer, 300 dpi, EU with cutter and unwinder
    1 * 1 KIT
    1 * 1 KIT - 724,00 €
    724,00 €
    1 * 1 KIT
       724,00 €
  • BBP™12 Desktop label printer, 300 dpi, EU-plug
    1 * 1 ST
    1 * 1 ST - 577,00 €
    577,00 €
    1 * 1 ST
       577,00 €
  • BBP™12 Desktop label printer, 300 dpi, LAB KIT, UK-plug
  • BBP™12 Desktop label printer, 300 dpi, with unwinder and LabelMark6 Pro software, UK-plug
  • BBP™12 Desktop label printer, LAB KIT and Brady workstation PWID (Product & Wire ID) suite, EU-plug
    1 * 1 KIT
    1 * 1 KIT - 1 620,00 €
    1 620,00 €
    1 * 1 KIT
       1 620,00 €
Accessories
  •  
    Description
    For
  • 116-0611
    External material holder
    BBP11/BBP12
    1 * 1 ST - 32,80 €
  •  
    Description
    For
  • BRDY149187
    Brady workstation automation software suite
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    1 * 1 ST - 833,00 €
  •  
    Description
  • BRDY195526
    Brady workstation CLP/GHS app via download
    1 * 1 ST - 369,99 €
  •  
    Description
    For
  • BRDY145445
    Workstation lockout writer software app
    BBP®31, BBP®33, BBP®85, i3300
    1 * 1 ST - 833,00 €
  •  
    Description
    For
  • BRDY149445
    Brady workstation scan and print software suite
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    1 * 1 ST - 833,00 €