ContactUsLinkComponent
Gas detection tubes for hydrochloric acid
Gas detection tubes for hydrochloric acid
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for hydrochloric acid
Supplier:  Dräger
Specifications
  • Catalog No:
  • 118-0328
  • 151-0152
  • 151-0203
  • Model:
  • Hydrochloric acid 0.2/a
  • Hydrochloric acid 50/a
  • Hydrochloric acid 1/a
  • Colour change:
  • Blue to yellow
  • Blue to pale yellow
  • Blue to yellow
  • Humidity:
  • ≤ 15 mg/l H₂O
  • ≤ 15 mg/l H₂O
  • ≤ 15 mg/l H₂O
  • Measurement time:
  • 2 min/25 s
  • 30 seconds/5 minutes
  • 2 min
  • No. of strokes:
  • 10/2
  • 1/10
  • 10
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 0,2 - 3/3 - 20 ppm
  • 500 - 5000/50 - 500 ppm
  • 1 - 10 ppm
  • Temperature range:
  • 5…40 °C
  • 10…50 °C
  • 5…40 °C