ContactUsLinkComponent
Gas detection tubes for water vapour
 :  Dräger
undefined
Gas detection tubes for water vapour
 :  Dräger
  • Catalog No:
  • 151-0170
  • 151-0180
  • 151-3152
  • DRAE8103061
  • Model:
  • Water vapour 0.1/a
  • Water vapour 1/b
  • Water vapour 0.1
  • Colour change:
  • Yellow to blue
  • Yellow to turquoise blue
  • Yellow to red brown
  • Measurement time:
  • 1,5 minutes
  • 20/40 seconds
  • 2 minutes
  • 600/300 seconds
  • No. of strokes:
  • 3
  • 1/2
  • 10
  • Standard deviation:
  • ±15 - 20 %
  • ±15 - 20 %
  • ±10 - 15 %
  • Standard range:
  • 0,1 - 1,0 mg/l
  • 20 - 40/1 - 18 mg/l
  • 1 - 40 mg/l
  • Temperature range:
  • 0…30 °C
  • 0…50 °C
  • 0…40 °C
Frequently Bought Together