Order Entry
Belgium
ContactUsLinkComponent
 
Closures for crimp neck vials 11 mm
  VARICP10048
 :  VARIAN
Closures for crimp neck vials 11 mm
  VARICP10048
 :  VARIAN

 

  • For:
    1 ml vials
  • Description:
    Aluminium caps, 11 mm
  • Pk:
    1.000