ContactUsLinkComponent
Gas detection tubes for chlorine
Gas detection tubes for chlorine
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for chlorine
Supplier:  Dräger
Certificates
Specifications
  • Catalog No:
  • 151-0191
  • 151-0195
  • Model:
  • Chlorine 50/a
  • Chlorine 0.2/a
  • Colour change:
  • Green grey to orange brown
  • White to yellow orange
  • Humidity:
  • ≤ 40 mg/l H₂O
  • ≤ 15 mg/l H₂O
  • Measurement time:
  • 20 seconds
  • 3 minutes/20 seconds
  • No. of strokes:
  • 1
  • 10/1
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 50 - 500 ppm
  • 0,2 - 3/3 - 30 ppm
  • Temperature range:
  • 10…40 °C
  • 10…40 °C