ContactUsLinkComponent
Gas detection tubes for sulphur dioxide
Gas detection tubes for sulphur dioxide
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for sulphur dioxide
Supplier:  Dräger
Certificates
Specifications
  • Catalog No:
  • 118-0308
  • 151-0146
  • 151-0207
  • 151-0194
  • 151-0154
  • Model:
  • Sulphur dioxide 50/b
  • Sulphur dioxide 0.1/a
  • Sulphur dioxide 1/a
  • Sulphur dioxide 20/a
  • Sulphur dioxide 0.5/a
  • Colour change:
  • Blue to yellow
  • Yellow to orange
  • Grey blue to white
  • Brown yellow to white
  • Grey blue to white
  • Humidity:
  • 1 -15 mg/l H₂O
  • 3 -15 mg/l H₂O
  • 3 -20 mg/l H₂O
  • ≤ 30 mg/l H₂O
  • ≤ 20 mg/l H₂O
  • Measurement time:
  • 15 seconds/3 minutes
  • 20 minutes
  • 3 minutes
  • 3 minutes
  • 3/6 minutes
  • No. of strokes:
  • 1/10
  • 100
  • 10
  • 10
  • 10/20
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 400 - 8000 / 50 - 500 ppm
  • 0,1 - 3 ppm
  • 1 - 25 ppm
  • 20 - 200 ppm
  • 1 - 25/0,5 - 5 ppm
  • Temperature range:
  • 0…50 °C
  • 10…30 °C
  • 15…25 °C
  • 0…40 °C
  • 15…30 °C