Orders LinkContactUsLinkComponent
Urinary system model
Urinary system model
Marketsource
Supplier:   3B Scientific
Urinary system model
Supplier:  3B Scientific
Marketsource
Certificates
Urinary system model
Urinary system model
2 Options Available Below
Product Details & Documents
Urinary system all-in-one model on baseboard showing:
  • Structures of retroperitoneal cavity
  • Large and small pelvis with bones and muscles
  • Inferior vena cava
  • Aorta with its branches including iliacal vessels
  • Upper urinary tract
  • Rectum
  • Kidney with adrenal gland
The prostate model provides a cross section of the male genital organs showing a healthy prostate with bladder, urethra, testicle, symphysis and rectum.
Product Family Options
Product Information
  • Availability
    Price
  • BINH1000319
    Prostate Model, 1/2 natural size
    Prostate Model, 1/2 natural size
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,00 €
    68,00 €
    1 * 1 ST
       68,00 €
    Specifications:
    Marketsource
    Marketsource
  • BINH1000317
    Dual Sex Urinary System, 6 part
    Dual Sex Urinary System, 6 part
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 472,00 €
    472,00 €
    1 * 1 ST
       472,00 €
    Specifications:
    Marketsource
    Marketsource