Orders LinkContactUsLinkComponent
Human cell model
Human cell model
Human cell in a glass
Catalog # BINH1008554
Marketsource
Supplier:   3B Scientific
Human cell model
Catalog # BINH1008554
Supplier:  3B Scientific
Marketsource
Human cell model
Human cell model
Human cell in a glass
Price
3 853,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This one of a kind model represents an undifferentiated human cell at an enlargement of 40000 times. It shows the structure of the smallest unit of any living creature capable of independent life, as seen through an electron microscope. The high quality human cell model shows the essential function-bearing cell organelles with stunning beauty and their arrangement in the model provides a momentary snapshot of the dynamic balance of a cell. The cell nucleus, a few mitochondria and the lysosomes are shown in section, so that their internal structure is visible. The glass cell is an eye-catcher in many exhibitions and has received several distinctions as 'World Didac Gold Award'.
  • Museum quality
  • 40000 times life size
Specifications
  • Description:
    Human cell in a glass