Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Formic acid
Formic acid ≥85%
Formic acid ≥85%
Formic acid ≥85%
APPC121029.1611
:
PanReac AppliChem
:
Formic acid ≥85%
APPC121029.1611
:
PanReac AppliChem
:
Pk:
1.000 mL
Seq:
2865
:
:
46.03 g/mol
:
13,04265
:
Ambient
:
200-579-1
:
1779
:
8,II
:
GHS05
:
64-18-6
:
HCO₂H
:
MFCD00003297