ContactUsLinkComponent
Gas detection tubes for carbon disulphide
Gas detection tubes for carbon disulphide
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for carbon disulphide
Supplier:  Dräger
Specifications
  • Catalog No:
  • 151-0183
  • DRAE6728351
  • 118-0339
  • Model:
  • Carbon disiphide 3/a
  • Carbon disiphide 5/a
  • Carbon disiphide 30/a
  • Colour change:
  • Pale blue to yellow green
  • White to brown green
  • Pale blue to brown
  • Measurement time:
  • 2 minutes
  • 3 minutes
  • 1 minute
  • No. of strokes:
  • 15 - 1
  • 11
  • 6
  • Standard deviation:
  • ±30%
  • ±10 - 15 %
  • ±15 - 20 %
  • Standard range:
  • 3 - 95 ppm
  • 5 - 60 ppm
  • 0,1 - 10 mg/l