Order Entry
Belgium
ContactUsLinkComponent
Anti-Cullin 1 Rabbit Polyclonal Antibody
Anti-Cullin 1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9542
Supplier:  Boster Bio
undefined
Anti-Cullin 1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9542
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cullin 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
     8454
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Cullin-1(CUL1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: Cullin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat