Orders LinkContactUsLinkComponent
Spray chamber
Spray chamber
Spray chamber
Catalog # HEWLG1820-65337
Supplier:   Agilent
Spray chamber
Catalog # HEWLG1820-65337
Supplier:  Agilent
Spray chamber
Spray chamber
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
This spray chamber made of quartz is used with 7500 ICP-MS.
It is dedicated for babington, concentric and micro-concentric nebulisers.
Caution: Not suitable for hydrofluoric acid.
Specifications
  • Description:
    Spray chamber