Orders LinkContactUsLinkComponent
Solvent filter/degasser
Solvent filter/degasser
Supplier:   Agilent
Solvent filter/degasser
Supplier:  Agilent
Certificates
Solvent filter/degasser
Solvent filter/degasser
7 Options Available Below
Product Details & Documents
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
  • Degasses eluents as particulates are removed
  • Increases solvent inlet lifetime
  • Eliminates pump downtime caused by air locks and particulates in check valves
  • Decreases piston wear, while increasing column life
Product Family Options
Product Information
  • Material
    Pore size
    Ø
    Availability
    Price
  • HEWL5188-2746
    Funnel base, glass
    Funnel base, glass
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL5188-2743
    Funnel, glass, 250 ml for HEWL3150-0577
    Funnel, glass, 250 ml for HEWL3150-0577
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL3150-0577
    HPLC solvent filter/degasser assembly
    HPLC solvent filter/degasser assembly
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL5188-2744
    PTFE coated sieve
    PTFE coated sieve
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL5188-2745
    PTFE seal for HEWL3150-0577
    PTFE seal for HEWL3150-0577
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Replacement filters
Product Information
  • Material
    Pore size
    Ø
    Availability
    Price
  • HEWL3150-0576
    RC filter membrane
    RC filter membrane
    0,45 μm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL3150-0509
    PTFE filter membrane
    PTFE filter membrane
    0,45 μm
    47 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST