Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forceps

Forceps

Straight, rounded tip (0,8 mm radius), White, 127 mm
Catalog # ENTEC01-0612
Supplier:   ENTEGRIS
Forceps
Catalog # ENTEC01-0612
Supplier:  ENTEGRIS
Forceps

Straight, rounded tip (0,8 mm radius), White, 127 mm

Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents

PP, white. Straight forceps for substrate handling.

 : Packaged in protective cases.

Specifications
  • Type:
    Straight, rounded tip (0,8 mm radius)
  • Colour:
    White
  • Length:
    127 mm
  • Seq:
    03