ContactUsLinkComponent
Mask packages, H 92 series
Mask packages, H 92 series
Catalog # ENTEH9260251061116
Supplier:  ENTEGRIS
Mask packages, H 92 series
Catalog # ENTEH9260251061116
Supplier:  ENTEGRIS
Specifications
  • Description:
    Mask size: 152×152 mm, thickness: 6,4 mm, overall size: 191×179×22 mm, non space for pellicles, material: PC, clear
  • Seq:
    08
  • Pk:
    12
Specifications
About this item
Provides excellent security for masks or reticles with pellicles.
  • High impact resistance
  • Stackable
  • Easy to open and close
  • Seal securely to provide a protective environment inside
  • Label wall for identification label