ContactUsLinkComponent
Mobile phase inlet filter, Hastelloy® C
Mobile phase inlet filter, Hastelloy® C
  VARIAL9402
 :  VARIAN
Mobile phase inlet filter, Hastelloy® C
  VARIAL9402
 :  VARIAN
  • Porosity:
    2 µm
  • Description:
    Mobile phase inlet filter with 1/8" tubing
  • Pk:
    1
This mobile phase inlet filter operates up to 500 °C and is compatible with halogenated solvents.