ContactUsLinkComponent
Shelving systems, stem castor cart with solid shelves, Super Erecta®
Shelving systems, stem castor cart with solid shelves, Super Erecta®
Catalog # METOF556EG
undefined
Shelving systems, stem castor cart with solid shelves, Super Erecta®
Catalog # METOF556EG
Specifications
  • Description:
    Stem castor cart with solid galvanised flat four shelves and 1600 mm chrome plated posts
  • W×D×H:
    1219×610×1724 mm
  • Pk:
    1
Specifications
About this item
The system consists of four solid shelves, posts, plastic split sleeves, donut bumpers and 127 mm poly castors (2 swivel; 2 brake). The raised 'ship's edge' around perimeter helps to contain spillage.
  • Chrome-plated posts with four galvanised flat shelves