ContactUsLinkComponent
Pneumatically operated diaphragm valves, Integra®
Pneumatically operated diaphragm valves, Integra®
Catalog # ENTE202-82-01
Supplier:  ENTEGRIS
Pneumatically operated diaphragm valves, Integra®
Catalog # ENTE202-82-01
Supplier:  ENTEGRIS
Specifications
  • Description:
    Valve type: normally closed, port connection 1/2" Flaretek® flared tube, PVDF nut
  • Pk:
    1
Specifications
About this item
Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution systems. Allows continuous flow through parallel ports. When actuated, the sample port opens allowing fluid to flow out or in. All wetted parts are molded PFA providing chemical resistance suitable for use in high purity, high corrosive chemical handling applications.
  • No external metal parts
  • Unique top loading design allows for field service
  • Piston shaft is encapsulated in PFA diaphragm to isolate it from corrosive fluids
  • Unique body design allows for 100% cavity drainage to prevent entrapment of fluids
  • Addition of Espy® sensors allows electronic monitoring of valve position
¹/₄" orifice, sampling, 3-way, position indication option.