PFA housing heads and bowls, for cartridge filters, Chemlock®
Supplier: ENTEGRIS
New Product
Certificates
About this item
PFA cartridge filter housing is the compact design, space-saving, enabes hassle-free cartridge changeouts (for safety and cleanliness) and reduced exposure to chemicals. It consists of lock-in design which perfectly seats the cartridge in the filter housing.
- Smaller footprint, minimal clearance cartridge/bowl design
- Cartridge locks into bowl with simple ¹/₄-turn twist
- Compact head with moulded on fittings
Operating temperature (max.): 100 °C
Operating pressure (max.): 43 psi at 100 °C and 110 psi at 25 °C
Frequently Bought Together